Merck
Brentuximab vedotin: first-line agent for advanced Hodgkin lymphoma.
Anticancer research (2013-09-12)
Huynh Cao, Kenneth Yamamoto, Li-Xi Yang, Robert Weber
ABSTRACT
Hodgkin lymphoma (HL) is characterized by malignant Reed-Sternberg cells which express CD30. Current National Comprehensive Cancer Network guidelines for patients with advanced HL (stage III/IV disease) recommend adriamycin, bleomycin, vinblastine, and dacarbazine (ABVD), or escalated bleomycin, etoposide, adriamycin, cyclophosphamide, vincristine, procarbazine, and prednisone (BEACOPP) as first-line regimens. ABVD appears to be as effective, with fewer side effects, as escalated BEACOPP. Escalated BEACOPP leads to a greater progression-free survival but no difference in overall survival. Recent advancements in technology have enabled an exciting shift to molecular-targeted cancer therapy. Brentuximab vedotin, a CD30-directed antibody conjugate, specifically targets malignant HL cells. It is approved by the Food and Drug Administration for the treatment of systemic anaplastic large-cell lymphoma and refractory HL that has progressed after autologous stem cell transplant, or after two prior multiagent chemotherapy regimens among patients ineligible to receive a transplant.
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg