Merck
  • Intake of coffee, caffeine and other methylxanthines and risk of Type I vs Type II endometrial cancer.
Intake of coffee, caffeine and other methylxanthines and risk of Type I vs Type II endometrial cancer.
British journal of cancer (2013-09-12)
S Uccella, A Mariani, A H Wang, R A Vierkant, W A Cliby, K Robien, K E Anderson, J R Cerhan
ABSTRACT
Coffee and other sources of methylxanthines and risk of Type I vs Type II endometrial cancer (EC) have not been evaluated previously. Prospective cohort of 23,356 postmenopausal women with 471 Type I and 71 Type II EC cases. Type I EC was statistically significantly associated with caffeinated (relative risk (RR)=0.65 for 4+ cups per day vs ≤1 cup per month: 95% confidence interval (CI): 0.47-0.89) but not decaffeinated (RR=0.76; 95% CI: 0.50-1.15) coffee intake; there were no associations with tea, cola or chocolate, or for Type II EC. The inverse association with caffeinated coffee intake was specific to women with a body mass index 30+ kg m(-2) (RR=0.56; 95% CI: 0.36-0.89). Coffee may protect against Type I EC in obese postmenopausal women.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard