Merck
  • The influence of amylose-LPC complex formation on the susceptibility of wheat starch to amylase.
The influence of amylose-LPC complex formation on the susceptibility of wheat starch to amylase.
Carbohydrate polymers (2013-08-06)
S Ahmadi-Abhari, A J J Woortman, A A C M Oudhuis, R J Hamer, K Loos
ABSTRACT
This study was aimed to assess the role of lysophosphatidylcholine (LPC) in the development of slowly digestible starch (SDS). The influence of LPC, on the enzymatic degradation of diluted 9% wheat starch suspensions (w/w) was investigated, using an in vitro digestion method. Wheat starch suspensions containing 0.5-5% LPC (based on starch) were heated in a Rapid Visco Analyser (RVA) till 95 °C and subjected to enzyme hydrolysis by porcine pancreatic α-amylase at 37 °C for several digestion periods. In vitro digestion measurements demonstrated that complexing starch with 5% LPC leads to a 22% decrease in rate of reducing sugar compared to the reference while the samples containing 0.5% LPC showed an equal digestibility comparable to the control. A clear decrease in the formation of reducing sugars was observed in presence of 2-5% LPC, since the results after 15 min digestion imply the formation of SDS due to the formation of amylose-LPC inclusion complexes. The DSC measurements proved the presence of amylose-LPC inclusion complexes even after 240 min digestion demonstrating the low susceptibility of amylose-V complexes to amylase.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein