Merck
  • Usage test with palladium-coated earrings in patients with contact allergy to palladium and nickel.
Usage test with palladium-coated earrings in patients with contact allergy to palladium and nickel.
Contact dermatitis (2013-08-06)
Cecilia Tillman, Malin Engfeldt, Monica Hindsén, Magnus Bruze
ABSTRACT
Since the EU Nickel Directive came into force, concern has been raised that palladium may increasingly replace nickel in alloys used in jewellery and dentistry, and that it may cause as many allergy problems as nickel. (i) To investigate the clinical relevance of contact allergy to palladium resulting from the wearing of palladium-coated earrings, and (ii) to evaluate the concordance in test results between palladium chloride (PdCl2 ), sodium tetrachloropalladate (Na2 PdCl4 ), and nickel(II) sulfate hexahydrate (NiSO4 .6H2 O), as well as variability in patch test reactivity. Forty female patients with pierced earlobes and previously known contact allergy to palladium and nickel were patch tested with dilution series of nickel and palladium salts. They wore earrings covered with palladium and titanium for 9 weeks to monitor the appearance of any clinical manifestations. There is a low risk of eczema in palladium-allergic and nickel-allergic individuals when they wear jewellery coated with pure palladium, but further studies are needed to determine whether palladium, when present in alloys, could cause allergic reactions. PdCl2 , like NiSO4 .6H2 O, shows variability in patch test reactivity over time. Furthermore, Na2 PdCl4 seems to be a more sensitive test substance than PdCl2 for the detection of palladium allergy.
Sigma-Aldrich
Nickel(II) sulfate heptahydrate, purum p.a., crystallized, ≥99.0% (KT)
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Sigma-Aldrich
Nickel(II) sulfate heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Sigma-Aldrich
Nickel(II) sulfate hexa-/ heptahydrate, DIN 50970 H, ≥20.6% Ni and Co basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) sulfate, anhydrous, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) sulfate hexahydrate, ACS reagent, ≥98%
Supelco
Nickel(II) sulfate hexahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel(II) sulfate hexahydrate, ReagentPlus®, powder or crystals