Merck
  • Geographic and temporal trends in antimicrobial nonsusceptibility in Streptococcus pneumoniae in the post-vaccine era in the United States.
Geographic and temporal trends in antimicrobial nonsusceptibility in Streptococcus pneumoniae in the post-vaccine era in the United States.
The Journal of infectious diseases (2013-07-16)
Ruth Link-Gelles, Ann Thomas, Ruth Lynfield, Sue Petit, William Schaffner, Lee Harrison, Monica M Farley, Deborah Aragon, Megin Nicols, Pam Daily Kirley, Shelley Zansky, James Jorgensen, Billie Anne Juni, Delois Jackson, Matthew R Moore, Marc Lipsitch
ABSTRACT
We examined whether observed increases in antibiotic nonsusceptible nonvaccine serotypes after introduction of pneumococcal conjugate vaccine in the United States in 2000 were driven primarily by vaccine or antibiotic use. Using active surveillance data, we evaluated geographic and temporal differences in serotype distribution and within-serotype differences during 2000-2009. We compared nonsusceptibility to penicillin and erythromycin by geography after standardizing differences across time, place, and serotype by regressing standardized versus crude proportions. A regression slope (RS) approaching zero indicates greater importance of the standardizing factor. Through 2000-2006, geographic differences in nonsusceptibility were better explained by within-serotype prevalence of nonsusceptibility (RS 0.32, 95% confidence interval [CI], .08-.55 for penicillin) than by geographic differences in serotype distribution (RS 0.71, 95% CI, .44-.97). From 2007-2009, serotype distribution differences became more important for penicillin (within-serotype RS 0.52, 95% CI, .11-.93; serotype distribution RS 0.57, 95% CI, .14-1.0). Differential nonsusceptibility, within individual serotypes, accounts for most geographic variation in nonsusceptibility, suggesting selective pressure from antibiotic use, rather than differences in serotype distribution, mainly determines nonsusceptibility patterns. Recent trends suggest geographic differences in serotype distribution may be affecting the prevalence of nonsusceptibility, possibly due to decreases in the number of nonsusceptible serotypes.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard