Merck
  • GABA protects human islet cells against the deleterious effects of immunosuppressive drugs and exerts immunoinhibitory effects alone.
GABA protects human islet cells against the deleterious effects of immunosuppressive drugs and exerts immunoinhibitory effects alone.
Transplantation (2013-07-16)
Gérald J Prud'homme, Yelena Glinka, Craig Hasilo, Steven Paraskevas, Xiaoming Li, Qinghua Wang
ABSTRACT
We recently found that γ-aminobutyric acid (GABA) protects mouse islet β cells. It prevented autoimmune type 1 diabetes in mice, induced islet β-cell regeneration, and exerted immunoinhibitory effects. However, it is not known whether GABA would be equally active on human islet and immune cells. In vitro culture of human islets and immune cells with or without GABA and immunosuppressive drugs. In vitro analysis of apoptosis, proliferation, nuclear factor (NF)-κB activation, calcium signaling, and insulin secretion. GABA reduced human islet cell apoptosis in culture, such that the yield of live cells was approximately tripled after 1 week, and it stimulated insulin secretion. It protected against the deleterious effects of rapamycin, tacrolimus, and mycophenolate mofetil. In human immune cells, GABA had inhibitory effects similar to mouse cells, such as suppressed anti-CD3-stimulated T-cell proliferation, in a GABA type A receptor-dependent fashion. The immunosuppressive mechanisms have been unclear, but we found that GABA blocked calcium influx, which is a key activation signal. GABA also suppressed NF-κB activation in both human islet cells and immune cells. We found that it could be combined with rapamycin to increase its suppressive effects. GABA improved human islet cell survival and had suppressive effects on human immune cells. It inhibited canonical NF-κB activation in both islet and immune cells. This is important because activation of this pathway is detrimental to islet cells and likely promotes damaging autoimmunity and alloreactivity against transplanted islets. These findings suggest that GABA might find applications in clinical islet transplantation.
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard