Merck
  • Cell response to PEGylated poly(dopamine) coated liposomes considering shear stress.
Cell response to PEGylated poly(dopamine) coated liposomes considering shear stress.
Biochimica et biophysica acta (2013-07-03)
Boon M Teo, Rebecca van der Westen, Leticia Hosta-Rigau, Brigitte Städler
ABSTRACT
Liposomes have gained immerse attention in the field of drug delivery as carriers of therapeutic molecules. Their modification with a polymer either to make them stealth (e.g. using PEG) and/or more stable (e.g. using poly(dopamine) (PDA)) is a crucial aspect to improve their performance e.g. the blood circulation time. Despite their potential, there are only a few commercialized liposome-based formulations for intravenous drug delivery. Hence, there is still considerable need to address the challenges involved in the design and characterization of liposomal therapeutics. In the latter case, it is of paramount importance to consider the dynamic in vivo environment, e.g. the interstitial fluidic pressure in tumors, blood flow, or bile flow in the liver. The PEGylation of PDA films was characterized by quartz crystal microbalance with dissipation monitoring, and the optimized protocol was used to assemble PEGylated PDA coated liposomes (LPDA_PEG). Dynamic light scattering, a plate reader, a flow cytometer and a cytotoxicity assay were used to characterize the liposomes and quantify cellular association/uptake and cell viability in the presence and absence of shear stress after 30min and 4h. The immortalized skeletal mouse myoblast (C2C12) cell line was chosen as model cancer cells, and a hepatic cell line (HepG2) was selected due to their importance in nanosized drug carrier clearance from the system in the liver. The presence of hydrophilic cargo did not affect the PDA assembly process. In the absence of shear stress, there was no difference in cellular uptake/association of both PDA coated liposomes (LPDA) and LPDA_PEG for hepatocytes while myoblasts preferentially internalized/associated with LPDA. In the presence of shear stress, hepatocytes preferentially internalized/associated with LPDA after 30min, while there was only a significant difference for myoblasts after 4h. The cell viability remained unaffected in all cases. LPDA_PEG are a promising platform towards drug delivery. The nature of cells and fluidic flow are important factors to be considered in their characterization using cell cultures. These findings will contribute in the better understanding of polymer coated liposomes with cells. The importance of microfluidics in cell culture based characterization is demonstrated, and this will eventually affect the way advanced drug delivery vehicles are designed and characterized prior to animal experiments.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 5,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Dextran, analytical standard, for GPC, 50,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, for GPC, 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, 12,000