Merck
  • Silver nanoparticle toxicity to Daphnia magna is a function of dissolved silver concentration.
Silver nanoparticle toxicity to Daphnia magna is a function of dissolved silver concentration.
Environmental toxicology and chemistry (2013-06-14)
Kim M Newton, Hema L Puppala, Christopher L Kitchens, Vicki L Colvin, Stephen J Klaine
ABSTRACT
The most persistent question regarding the toxicity of silver nanoparticles (AgNPs) is whether this toxicity is due to the nanoparticles themselves or the silver ions (Ag(+)) they release. The present study investigates the role of surface coating and the presence of dissolved organic carbon on the toxicity of AgNPs to Daphnia magna and tests the hypothesis that the acute toxicity of AgNPs is a function of dissolved Ag produced by nanoparticle dissolution. Toxicity of silver nitrate (AgNO3) and AgNPs with surface coatings-gum arabic (AgGA), polyethylene glycol (AgPEG), and polyvinylpyrrolidone (AgPVP)-at 48 h was assessed in US Environmental Protection Agency moderately hard reconstituted water alone and augmented with Suwannee River dissolved organic carbon (DOC). As expected, AgNO3 was the most toxic to D. magna and AgPVPs were the least toxic. In general, Suwannee River DOC presence reduced the toxicity of AgNO3, AgGAs, and AgPEG, while the toxicity of AgPVPs was unaffected. The measured dissolved Ag concentrations for all AgNPs and AgNO3 at the 48-h median lethal concentration in moderately hard reconstituted water were similar. The presence of Suwannee River DOC decreased the ratio of measured dissolved Ag to measured total Ag concentration. These results support the hypothesis that toxicity of AgNPs to D. magna is a function of dissolved Ag concentration from these particles.
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.