Merck
  • The cigarette smoke constituent benzo[a]pyrene disrupts metabolic enzyme, and apoptosis pathway member gene expression in ovarian follicles.
The cigarette smoke constituent benzo[a]pyrene disrupts metabolic enzyme, and apoptosis pathway member gene expression in ovarian follicles.
Reproductive toxicology (Elmsford, N.Y.) (2013-06-12)
Jean Clair Sadeu, Warren G Foster
ABSTRACT
Benzo[a]pyrene (B[a]P) is a prototypical polycyclic aromatic hydrocarbon (PAH) present in cigarette smoke. We previously showed that B[a]P adversely affects follicular development and survival. The objective of this study was to identify the key molecular pathways underlying B[a]P-induced abnormal follicular development. Isolated follicles (100-130 μm) from ovaries of F1 hybrid (C57BL/6j×CBA/Ca) mice were cultured for 8 (preantral/antral follicles) and 12 (preovulatory follicles) days in increasing concentrations of B[a]P (0 ng/mL [control] to 45 ng/mL). Expression of aryl hydrocarbon receptor (AhR), aryl hydroxylase steroidogenic enzyme, cell-cycle, and apoptotic genes were quantified. B[a]P exposure significantly (P<0.05) increased mRNA expression of Cyp1a1 in preantral/antral follicles and Cyp1b1, Bax and Hsp90ab1 in preovulatory follicles. No significant effect on mRNA expression of StAR, Cyp11a1, aromatase, Cdk4, Cdk2, Ccnd2, cIAP2, and survivin was observed. In conclusion, this study suggests that B[a]P exposure significantly affects the phase I enzymes and cell death genes during preantral/antral and preovulatory growth, and thus highlight the AhR signaling and apoptotis pathways in delayed follicle growth and decreased viability.
Product Description
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard