Merck
  • Mortadella sausage manufactured with Caiman yacare (Caiman crocodilus yacare) meat, pork backfat, and soybean oil.
Mortadella sausage manufactured with Caiman yacare (Caiman crocodilus yacare) meat, pork backfat, and soybean oil.
Meat science (2013-06-12)
C S N Morais, N N Morais Júnior, J Vicente-Neto, E M Ramos, J Almeida, C Roseiro, C Santos, L T Gama, M C Bressan
ABSTRACT
Mortadellas manufactured using Caiman yacare meat (70%) and pork fat (30%) were reformulated by substituting pork fat with increasing amounts of soybean oil (25%, 50%, and 100%) and evaluated 7, 30, and 60 days post-manufacture. The substitutions resulted in an increase in fat content and in the percentage of polyunsaturated fatty acids (total PUFA; 18:2 n-6 and 18:3 n-3), and a reduction in saturated fatty acids (total SFA; 14:0 and 16:0). These alterations reduced (P<0.01) the indices of atherogenicity and thrombogenicity by 4- to 5-fold in the formulation with 100% soybean oil compared to the original formulation. The levels of TBARS obtained at day 7 were positively correlated (P<0.01) with 18:2 n-6 and 18:3 n-3 (r=0.83 and 0.84, respectively) and negatively correlated with 17:0 and 17:1 (r=-0.91 and -0.89, respectively). All formulations received favourable overall acceptability by the sensory panel.