Merck
  • Sorption behavior of 17 phthalic acid esters on three soils: effects of pH and dissolved organic matter, sorption coefficient measurement and QSPR study.
Sorption behavior of 17 phthalic acid esters on three soils: effects of pH and dissolved organic matter, sorption coefficient measurement and QSPR study.
Chemosphere (2013-06-08)
Fen Yang, Meng Wang, Zunyao Wang
ABSTRACT
This work studies the sorption behaviors of phthalic acid esters (PAEs) on three soils by batch equilibration experiments and quantitative structure property relationship (QSPR) methodology. Firstly, the effects of soil type, dissolved organic matter and pH on the sorption of four PAEs (DMP, DEP, DAP, DBP) are investigated. The results indicate that the soil organic carbon content has a crucial influence on sorption progress. In addition, a negative correlation between pH values and the sorption capacities was found for these four PAEs. However, the effect of DOM on PAEs sorption may be more complicated. The sorption of four PAEs was promoted by low concentrations of DOM, while, in the case of high concentrations, the influence of DOM on the sorption was complicated. Then the organic carbon content normalized sorption coefficient (logKoc) values of 17 PAEs on three soils were measured, and the mean values ranged from 1.50 to 7.57. The logKoc values showed good correlation with the corresponding logKow values. Finally, two QSPR models were developed with 13 theoretical parameters to get reliable logKoc predictions. The leave-one-out cross validation (CV-LOO) indicated that the internal predictive power of the two models was satisfactory.
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard