Merck
  • Comparing the effects of nanosilver size and coating variations on bioavailability, internalization, and elimination, using Lumbriculus variegatus.
Comparing the effects of nanosilver size and coating variations on bioavailability, internalization, and elimination, using Lumbriculus variegatus.
Environmental toxicology and chemistry (2013-05-21)
Jessica G Coleman, Alan J Kennedy, Anthony J Bednar, James F Ranville, Jennifer G Laird, Ashley R Harmon, Charolett A Hayes, Evan P Gray, Christopher P Higgins, Guilherme Lotufo, Jeffery A Steevens
ABSTRACT
As the production and applications of silver nanoparticles (AgNPs) increase, it is essential to characterize fate and effects in environmental systems. Nanosilver materials may settle from suspension; therefore, the authors' objective was to utilize environmentally relevant bioassays and study the impact, bioaccumulation, tissue distribution, uptake, and depuration of AgNPs on a sediment-dwelling invertebrate, Lumbriculus variegatus. Hydrodynamic diameters of uncoated 30-nm, 80-nm, and 1500-nm AgNP powders and a polyvinyl pyrrolidone (PVP) AgNP suspension were measured utilizing dynamic light scattering in freshwater media (0-280 µS/cm). Aggregation for 30 nm, 80 nm, and 1500 nm silver increased with conductivity but was minimal for PVP silver. Lumbriculus variegatus were exposed to AgNPs or silver nitrate (AgNO3 ) spiked into sediment (nominally 100 mg/kg) and water (PVP 30 nm and 70 nm Ag, nominally 5 mg/L). Uptake was assessed through inductively coupled plasma mass spectroscopy (ICP-MS) and hyperspectral imaging. Particle sizes were examined through field flow fractionation-ICP-MS (FFF-ICP-MS) and ICP-MS in single particle mode (SP-ICP-MS). Lumbriculus variegatus were also depurated for 6 h, 8 h, 24 h, and 48 h to determine gut clearance. Bioaccumulation factors of sediment-exposed L. variegatus were similar regardless of particle size or coatings. The FFF-ICP-MS and SP-ICP-MS detected AgNPs for up to 48 h post depuration. The present study provides information on bioaccumulation and interactions of AgNPs within biological systems.
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.