Merck
  • Isolation of Clostridium difficile from faecal specimens--a comparison of chromID C. difficile agar and cycloserine-cefoxitin-fructose agar.
Isolation of Clostridium difficile from faecal specimens--a comparison of chromID C. difficile agar and cycloserine-cefoxitin-fructose agar.
Journal of medical microbiology (2013-04-13)
Kerry C Carson, Lusiana V Boseiwaqa, Sara K Thean, Niki F Foster, Thomas V Riley
ABSTRACT
The culture of toxigenic Clostridium difficile from stool specimens is still seen as the gold standard for the laboratory diagnosis of C. difficile infection (CDI). bioMérieux have released ChromID Cdiff chromogenic agar (CDIF) for the isolation and identification of C. difficile in 24 h. In this study, we compared CDIF to pre-reduced cycloserine-cefoxitin-fructose agar with sodium taurocholate (TCCFA) in the examination of glutamate dehydrogenase-positive faecal specimens that were either GeneOhm positive or negative, using direct culture or culture following alcohol shock. Direct culture on CDIF had a sensitivity of 100 % and recovery of 94 % while for TCCFA these were 87 % and 82 %, respectively. For GeneOhm-positive alcohol-shocked faecal samples, sensitivity and recovery on CDIF was similar to direct culture while on TCCFA they were about 10 % higher. For direct culture, there was a significant difference between growth on CDIF at 24 h and TCCFA at 48 h (P = 0.001) and between the two media at 48 h (P<0.001). A total of 142 strains of C. difficile were recovered in pure culture from all GeneOhm-positive samples used in this study and 11 (7.7 %) of these were A(-)B(-)CDT(-) and may represent mixed infections of toxigenic and non-toxigenic C. difficile. The most dominant ribotype was UK 014 (14.7 %) followed by 002 (11.9 %) and 020 (11.9 %), and 36 % of toxigenic isolates, including an A(-)B(+)CDT(-) strain, could not be assigned a UK ribotype. CDIF outperformed pre-reduced TCCFA by negating the need for alcohol shock treatment and by giving a time saving of 24 h in the isolation of C. difficile. CDIF plates were also more selective than TCCFA and C. difficile colonies were easy to identify and subculture prior to strain typing.
Product Description
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, ash 2.0-4.5%
Supelco
Cefoxitin sodium salt
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Cefoxitin sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
Bacteriological agar, for molecular biology
Sigma-Aldrich
D-Cycloserine, synthetic