Merck
  • Synthesis and activity of novel benzoxazole derivatives of mannopeptimycin glycopeptide antibiotics.
Synthesis and activity of novel benzoxazole derivatives of mannopeptimycin glycopeptide antibiotics.
Bioorganic & medicinal chemistry letters (2003-07-11)
Phaik-Eng Sum, David How, Nancy Torres, Howard Newman, Peter J Petersen, Tarek S Mansour
ABSTRACT
A series of benzoxazole derivatives of the mannopeptimycin glycopeptide antibiotics was synthesized via a novel benzoxazole formation reaction by treating aminophenol of mannopeptimycin-beta with an aldehyde and DDQ in DMF. Some of these derivatives (e.g., 5b, 5d, 5m, and 7b) showed good activity against Gram-(+) bacteria when compared to the parent compound mannopeptimycin-beta.