Merck
  • Vibrational spectra, assignments and normal coordinate analysis of 3-aminobenzyl alcohol.
Vibrational spectra, assignments and normal coordinate analysis of 3-aminobenzyl alcohol.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-09-10)
N Sundaraganesan, H Saleem, S Mohan
ABSTRACT
The laser Raman and FTIR spectra of 3-aminobenzyl alcohol have been recorded. The observed frequencies were assigned to various modes of vibrations on the basis of normal coordinate analysis, assuming C(s) point group symmetry. The potential energy distribution associated with normal modes is also reported here. The assignment of fundamental vibrations agrees well with the calculated frequencies.
Sigma-Aldrich
3-Aminobenzyl alcohol, 97%