Merck
  • Density functional theory study of Fourier transform infrared and Raman spectra of 2-amino-5-chloro benzonitrile.
Density functional theory study of Fourier transform infrared and Raman spectra of 2-amino-5-chloro benzonitrile.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-04-22)
V Krishnakumar, S Dheivamalar
ABSTRACT
The vibrational spectra of 2-amino-5-chloro benzonitrile (ACB) have been obtained by density functional theory (DFT) calculations. Normal coordinate analysis has been carried out to support the vibrational analysis. The results were compared with the experimental values. With the help of scaling procedures, the observed FTIR and FT Raman vibrational frequencies were analysed and compared with the theoretically predicted vibrational spectra. The assignments of bands to various normal modes of the molecules were also carried out.
Sigma-Aldrich
2-Amino-5-chlorobenzonitrile, 98%
Pricing and availability is not currently available.