Merck
  • Self-activating nuclease activity of copper (II) complexes of hydroxyl-rich ligands.
Self-activating nuclease activity of copper (II) complexes of hydroxyl-rich ligands.
Journal of inorganic biochemistry (2005-11-18)
Sunil S Tonde, Avinash S Kumbhar, Subhash B Padhye, Ray J Butcher
ABSTRACT
Copper (II) complexes of Schiff bases derived from [1+1] condensation of salicylaldehyde, 2,3-dihydroxybenzaldehyde and 2,3,4-trihydroxybenzaldehyde with anthranilic acid (L1-L3) have been synthesized and characterized by elemental analyses, IR, UV-Vis spectra, room temperature magnetic susceptibility, electron paramagnetic resonance spectroscopy and cyclic voltammetry. The X-ray structure of [CuL1]n has been solved and refined to R = 0.0314. The crystals are monoclinic with space group P2(1) with cell constants a = 9.6820(13), b = 7.1446(11), c = 9.9315(13) A, beta = 98.385(8) degrees, Z = 2. The copper (II) ions are in a distorted tetrahedral environment sequentially bridged by carboxylate groups in the syn-anti conformation giving rise to a helix-like chain. The copper complexes with the inherent redox active hydroquinone functionality cleave plasmid pBR322 DNA without exogenous agents by a self-activating mechanism.
Sigma-Aldrich
2,3-Dihydroxybenzaldehyde, 97%
Sigma-Aldrich
2,3,4-Trihydroxybenzaldehyde, 98%