- Activity-based near-infrared fluorescent probe for LMP7: a chemical proteomics tool for the immunoproteasome in living cells.
Activity-based near-infrared fluorescent probe for LMP7: a chemical proteomics tool for the immunoproteasome in living cells.
Chembiochem : a European journal of chemical biology (2012-07-19)
Lalit Kumar Sharma, Na-Ra Lee, Eun Ryoung Jang, Beilei Lei, Chang-Guo Zhan, Wooin Lee, Kyung-Bo Kim
PMID22807337
ABSTRACT
Probing the unknown: The immunoproteasome, an alternative form of the constitutive proteasome, has been implicated in a number of pathological states such as cancer and autoimmune diseases. In an effort to understand the role of the immunoproteasome in cells, the first immunoproteasome-specific near-infrared fluorescent probe has been developed.