Merck
Phase-transfer alkylation reactions using microreactors.
Chemical communications (Cambridge, England) (2003-05-15)
Masaharu Ueno, Hideaki Hisamoto, Takehiko Kitamorib, Shŭ Kobayashi
ABSTRACT
Phase-transfer alkylation in a microreactor proceeds smoothly, and the reaction has been found to be more efficient than that in a round-bottomed flask with vigorous stirring; we have observed by an optical microscope study that an interfacial area provided by organic and aqueous phases is more extended in a microreactor.
Sigma-Aldrich
Ethyl 2-oxocyclopentanecarboxylate, 95%
Pricing and availability is not currently available.