Merck
  • Versatile stereoselective cycloadditions between heterocumulenes and phosphagermaallene Tip(tBu)Ge=C=PMes*: experimental and theoretical investigations.
Versatile stereoselective cycloadditions between heterocumulenes and phosphagermaallene Tip(tBu)Ge=C=PMes*: experimental and theoretical investigations.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-10-01)
Dumitru Ghereg, Erwan André, Heinz Gornitzka, Jean Escudié, Fatima Ouhsaine, Nathalie Saffon, Karinne Miqueu, Jean-Marc Sotiropoulos
ABSTRACT
Phosphagermaallene Tip(tBu)Ge=C=PMes* 1 (Tip=2,4,6-triisopropylphenyl, Mes*=2,4,6-tri-tert-butylphenyl) reacts with phenyl isocyanate and tert-butyl isocyanate by a [2+2] cycloaddition that involves the Ge=C and C=O double bonds to afford 1-oxa-2-germacyclobutanes 2 and 3. With N,N'-dicyclohexylcarbodiimide, a [2+2] cycloaddition is observed between the Ge=C and C=N unsaturations to lead to 1-aza-2-germacyclobutane 6 with exocyclic P=C and C=N double bonds. In sharp contrast, 1 reacts with phenyl isothiocyanate, ethyl isothiocyanate, and carbon disulfide according to a [3+2] cycloaddition that involves the whole Ge=C=P unit and the C=S double bond to give transient phosphagermacarbenes (PGeHCs) 11, 12, and 13. These new PGeHCs undergo C-H insertions into one o-tBu group of Mes* (in the case of 11 and 12) or one o-iPr group of Tip (in the case of 13) with formation of tricyclic compounds 8, 9, and 10, respectively. The reaction mechanisms that involve 1 and the phenyl isocyanate and the phenyl isothiocyanate are described and their regioselectivity is explained by theoretical calculations.
Sigma-Aldrich
tert-Butyl isocyanate, 97%