Merck
  • Simple and automatic determination of aldehydes and acetone in water by headspace solid-phase microextraction and gas chromatography-mass spectrometry.
Simple and automatic determination of aldehydes and acetone in water by headspace solid-phase microextraction and gas chromatography-mass spectrometry.
Journal of separation science (2011-02-15)
Hyun-Ji Kim, Ho-Sang Shin
ABSTRACT
We describe a simple and automatic method to determine nine aldehydes and acetone simultaneously in water. This method is based on derivatization with 2,2,2-trifluoroethylhydrazine (TFEH) and consecutive headspace-solid-phase microextraction and gas chromatography-mass spectrometry. Acetone-d(6) was used as the internal standard. Aldehydes and acetone in water reacted for 30 min at 40°C with TFEH in a headspace vial and the formed TFEH derivatives were simultaneously vaporized and adsorbed on polydimethylsiloxane-divinylbenzene. Under the established condition, the method detection limit was 0.1-0.5 μg/L in 4 mL water and the relative standard deviation was less than 13% at concentrations of 0.25 and 0.05 mg/L. This method was applied to determine aldehydes and acetone in 5 mineral water and 114 surface water samples. All mineral water samples had detectable levels of methanal (24.0-61.8 μg/L), ethanal (57.7-110.9 μg/L), propanal (11.5-11.7 μg/L), butanal, pentanal (3.3-3.4 μg/L) and nonanal (0.3-0.4 μg/L). Methanal and ethanal were also detected in concentration range of 2.7-117.2 and 1.2-11.9 μg/L, respectively, in surface water of 114 monitoring sites in Korea.
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
2,2,2-Trifluoroethylhydrazine solution, 70 wt. % in H2O
Sigma-Aldrich
Acetone-d6, 99.9 atom % D