Merck
Inhibition of cancer cell growth by ruthenium(II) arene complexes.
Journal of medicinal chemistry (2001-10-19)
R E Morris, R E Aird, P del S Murdoch, H Chen, J Cummings, N D Hughes, S Parsons, A Parkin, G Boyd, D I Jodrell, P J Sadler
ABSTRACT
Inhibition of the growth of the human ovarian cancer cell line A2780 by organometallic ruthenium(II) complexes of the type [(eta(6)-arene)Ru(X)(Y)(Z)], where arene is benzene or substituted benzene, X, Y, and Z are halide, acetonitrile, or isonicotinamide, or X,Y is ethylenediamine (en) or N-ethylethylenediamine, has been investigated. The X-ray crystal structures of the complexes [(eta(6)-p-cymene)Ru(en)Cl]PF(6) (5), [(eta(6)-p-cymene)RuCl(2)(isonicotinamide)] (7), and [(eta(6)-biphenyl)Ru(en)Cl]PF(6) (9) are reported. They have "piano stool" geometries with eta(6) coordination of the arene ligand. Complexes with X,Y as a chelated en ligand and Z as a monofunctional leaving group had the highest activity. Complexes 5, 6 (the iodo analogue of 5), 9, and 10 (ethylethylenediamine analogue of 9) were as active as carboplatin. Hydrolysis of the reactive Ru-Cl bond in complex 5 was detected by HPLC but was suppressed by the addition of chloride ions. Complex 5 binds strongly and selectively to G bases on DNA oligonucleotides to form monofunctional adducts. No inhibition of topoisomerase I or II by complexes 5, 6, or 9 was detected. These chelated Ru(II) arene complexes have potential as novel metal-based anticancer agents with a mechanism of action different from that of the Ru(III) complex currently on clinical trial.
Sigma-Aldrich
N-Ethylethylenediamine, 98%
Pricing and availability is not currently available.