Merck
Synthesis of oligo-RNAs with photocaged adenosine 2'-hydroxyls.
Nature protocols (2007-06-05)
Steven G Chaulk, Andrew M MacMillan
ABSTRACT
This protocol describes a general method for the preparation of RNAs in which the reactivity or hydrogen-bonding properties of the molecule are modified in a photoreversible fashion by use of a caging strategy. A single caged adenosine, modified at the 2' position as a nitro-benzyl ether, can be incorporated into short RNAs by chemical synthesis or into long RNAs by a combination of chemical and enzymatic synthesis. The modified RNAs can be uncaged by photolysis under a variety of conditions including the use of a laser or xenon lamp, and the course of this uncaging reaction may be readily followed by HPLC or thin-layer chromatography.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent