Merck
  • Increased sensitivity and specificity of Borrelia burgdorferi 16S ribosomal DNA detection.
Increased sensitivity and specificity of Borrelia burgdorferi 16S ribosomal DNA detection.
American journal of clinical pathology (2010-03-17)
Sin Hang Lee, Veronica S Vigliotti, Jessica S Vigliotti, William Jones, Suri Pappu
ABSTRACT
The DNA of Borrelia burgdorferi spirochetes extracted by ammonium hydroxide was used as the template for nested polymerase chain reaction (PCR) amplification of the species-specific 16S ribosomal DNA (rDNA). The primers were those well known to be specific for signature sequence amplification of the B burgdorferi sensu lato 16S ribosomal RNA gene. The positive 293-base-pair nested PCR amplicon was subjected to routine direct automated Sanger sequencing. A 50-base sequence excised randomly from the sequencing electrophoretogram between the 2 nested PCR primer binding sites was sufficient for the Basic Local Alignment Search Tool (BLAST) analysis to validate the B burgdorferi sensu lato 16S rDNA without a reasonable doubt. Nested PCR increased the sensitivity of DNA detection by 100- to 1,000-fold. DNA sequence validation based on BLAST algorithms using the GenBank database practically eliminates any possibility of false-positive results due to molecular misidentification. This technology may be a valuable supplement to the current serologic tests for Lyme disease.
Sigma-Aldrich
BSK-H Medium, Complete, sterile-filtered, with 6% rabbit serum, suitable for Borrelia burgdorferi
Pricing and availability is not currently available.