Merck
  • Enrichment of mutations in multiple DNA sequences using COLD-PCR in emulsion.
Enrichment of mutations in multiple DNA sequences using COLD-PCR in emulsion.
PloS one (2012-12-14)
Elena Castellanos-Rizaldos, Coren Audrey Milbury, G Mike Makrigiorgos
ABSTRACT
Multiplex detection of low-level mutant alleles in the presence of wild-type DNA would be useful for several fields of medicine including cancer, pre-natal diagnosis and infectious diseases. COLD-PCR is a recently developed method that enriches low-level mutations during PCR cycling, thus enhancing downstream detection without the need for special reagents or equipment. The approach relies on the differential denaturation of DNA strands which contain Tm-lowering mutations or mismatches, versus 'homo-duplex' wild-type DNA. Enabling multiplex-COLD-PCR that can enrich mutations in several amplicons simultaneously is desirable but technically difficult to accomplish. Here we describe the proof of principle of an emulsion-PCR based approach that demonstrates the feasibility of multiplexed-COLD-PCR within a single tube, using commercially available mutated cell lines. This method works best with short amplicons; therefore, it could potentially be used on highly fragmented samples obtained from biological material or FFPE specimens. Following a multiplex pre-amplification of TP53 exons from genomic DNA, emulsions which incorporate the multiplex product, PCR reagents and primers specific for a given TP53 exon are prepared. Emulsions with different TP53 targets are then combined in a single tube and a fast-COLD-PCR program that gradually ramps up the denaturation temperature over several PCR cycles is applied (temperature-tolerant, TT-fast-eCOLD-PCR). The range of denaturation temperatures applied encompasses the critical denaturation temperature (T(c)) corresponding to all the amplicons included in the reaction, resulting to a gradual enrichment of mutations within all amplicons encompassed by emulsion. Validation for TT-fast-eCOLD-PCR is provided for TP53 exons 6-9. Using dilutions of mutated cell-line into wild-type DNA, we demonstrate simultaneous mutation enrichment between 7 to 15-fold in all amplicons examined. TT-fast-eCOLD-PCR expands the versatility of COLD-PCR and enables high-throughput enrichment of low-level mutant alleles over multiple sequences in a single tube.
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Span® 80, nonionic surfactant
Sigma-Aldrich
TWEEN® 80, viscous liquid