Merck
  • Poly(dG)-poly(dC) sequences, under torsional stress, induce an altered DNA conformation upon neighboring DNA sequences.
Poly(dG)-poly(dC) sequences, under torsional stress, induce an altered DNA conformation upon neighboring DNA sequences.
Cell (1985-11-01)
T Kohwi-Shigematsu, Y Kohwi
ABSTRACT
Supercoiled plasmid DNAs (at bacterial superhelical density) harboring the homopurine-homopyrimidine sequence, poly(dG)-poly(dC), were reacted with bromoacetaldehyde (BAA), a reagent that reacts with unpaired DNA bases. Not only did the poly(dG)-poly(dC) sequence react with BAA but, surprisingly, neighboring sequences located 3' to the contiguous G sequences also reacted. The altered conformation in the poly(dG)-poly(dC) sequence and in the neighboring sequence occurred in the same supercoiled plasmid DNA molecule. Furthermore, the occurrence of an "unpaired" conformation in the neighboring sequence is strictly due to a positional effect, since it is observed when the poly(dG)-poly(dC) segment is adjacent to a variety of neighboring sequences.
Sigma-Aldrich
Poly(2′-deoxyguanyl-2′-deoxycytidylic acid) sodium salt, ampule of >10 A260