Merck
  • Presence of N-methyl-norsalsolinol in the CSF: correlations with dopamine metabolites of patients with Parkinson's disease.
Presence of N-methyl-norsalsolinol in the CSF: correlations with dopamine metabolites of patients with Parkinson's disease.
Journal of the neurological sciences (1995-08-01)
A Moser, J Scholz, F Nobbe, P Vieregge, V Böhme, H Bamberg
ABSTRACT
We could identify the MPTP-like compound and isoquinoline derivative N-methyl-norsalsolinol (2-MDTIQ) in cerebrospinal fluid (CSF) of patients with Parkinson's disease. The presence of 2-MDTIQ negatively correlated with the disease duration. In order to study the relationship between presence of 2-MDTIQ and dopamine metabolism, we examined 3-O-methyl-dopa (MDOPA) and homovanillic acid (HVA) levels in CSF of 15 normal control subjects and 34 patients with Parkinson's disease (PD). In the PD group in which 2-MDTIQ was detected, the HVA/MDOPA ratio was also negatively correlated with the duration of the disease and was increased when compared to patients without 2-MDTIQ. Since in both PD groups the daily L-dopa dose, the mean MDOPA levels, and the daily L-dopa dose/MDOPA ratio were nearly identical the results are not related to different L-dopa medications. In vitro experiments demonstrated 2-MDTIQ to inhibit monoamine oxidase activity in the caudate-putamen. These results suggest that 2-MDTIQ indicates an increased dopamine turnover in patients with PD. The enhanced metabolism at the beginning of the disease is not due to the presence of 2-MDTIQ since it inhibits dopamine metabolism. Thus, 2-MDTIQ, probably endogenously synthesized from dopamine, appears as a result of a compensatively activated dopaminergic system.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Tin, ≥99%, powder
SAFC
L-Cystine
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis