Merck
  • Plasma 3-methylbutanal in man and its relationship to hepatic encephalopathy.
Plasma 3-methylbutanal in man and its relationship to hepatic encephalopathy.
Clinical physiology (Oxford, England) (1985-02-01)
A W Marshall, M DeSouza, M Y Morgan
ABSTRACT
3-Methylbutanal is a volatile aldehyde which in the rat is derived, at least in part, from colonic bacterial breakdown of leucine. It has been proposed as a toxin of importance in the pathogenesis of hepatic encephalopathy in man. A rapid, reliable and highly reproducible method for estimating 3-methylbutanal in plasma is described using a gas-chromatograph with a head space-sampler. The mean plasma 3-methylbutanal concentration in non-fasting patients with hepatic encephalopathy, 0.244 mumol/litre (range 0-1.30) was not significantly different from the mean value in controls, 0.116 mumol/litre (0-0.349). Oral leucine feeding resulted in significant increases in plasma 3-methylbutanal concentrations in both control subjects and patients with cirrhosis. Peak leucine and 3-methylbutanal values occurred at approximately the same time and usually within 120 min of leucine ingestion. Pre-treatment with neomycin had no effect on the results of leucine feeding. No changes occurred in the clinical condition or psychometric performance of patients with cirrhosis fed leucine despite increases in plasma 3-methylbutanal of up to 700% over basal values. In man plasma 3-methylbutanal, at least in part, derives from ingested leucine independently of the action of colonic bacteria. The role of this compound in the pathogenesis of hepatic encephalopathy in man needs further exploration.
Sigma-Aldrich
Isovaleraldehyde, natural, ≥95%, FG
Sigma-Aldrich
Isovaleraldehyde, ≥97%, FG
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Isovaleraldehyde, 97%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG
Sigma-Aldrich
2-Heptanone, 99%