Merck
Plasma leucine enkephalin is increased in liver disease.
Gut (1989-10-01)
J R Thornton, M S Losowsky
ABSTRACT
Plasma methionine enkephalin is increased in liver disease and may contribute to some of the clinical manifestations of hepatic failure. To determine if another 'small' opioid peptide is increased in the plasma of patients with liver disease, leucine enkephalin was measured by radioimmunoassay. Its plasma concentration was raised approximately five-fold in patients with acute liver disease (median 1490 pmol/l, range 830-2420) and three-fold in patients with cirrhosis with ascites (960 pmol/l, 470-2900), compared with disease controls (325 pmol/l, 180-740) and healthy controls (305 pmol/l, 180-560). The increase in plasma leucine enkephalin was proportional to the degree of liver damage, as judged in the patients with acute liver disease by its correlation with the prothrombin time (r = 0.691, p less than 0.01) and alanine aminotransferase (r = 0.502, p less than 0.05), and in the patients with cirrhosis by its negative correlation with the plasma albumin (r = -0.743, p less than 0.001). It is unclear whether the raised plasma leucine enkephalin in liver disease is a consequence of diminished hepatic inactivation, increased secretion from sympathetic nerves and adrenal glands, or both.
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)