Merck
  • Increases in cerebrospinal fluid caffeine concentration are associated with favorable outcome after severe traumatic brain injury in humans.
Increases in cerebrospinal fluid caffeine concentration are associated with favorable outcome after severe traumatic brain injury in humans.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2007-08-09)
Kathleen T Sachse, Edwin K Jackson, Stephen R Wisniewski, Delbert G Gillespie, Ava M Puccio, Robert S B Clark, C Edward Dixon, Patrick M Kochanek
ABSTRACT
Caffeine, the most widely consumed psychoactive drug and a weak adenosine receptor antagonist, can be neuroprotective or neurotoxic depending on the experimental model or neurologic disorder. However, its contribution to pathophysiology and outcome in traumatic brain injury (TBI) in humans is undefined. We assessed serial cerebrospinal fluid (CSF) concentrations of caffeine and its metabolites (theobromine, paraxanthine, and theophylline) by high-pressure liquid chromatography/ultraviolet in 97 ventricular CSF samples from an established bank, from 30 adults with severe TBI. We prospectively selected a threshold caffeine level of > or = 1 micromol/L (194 ng/mL) as clinically significant. Demographics, Glasgow Coma Scale (GCS) score, admission blood alcohol level, and 6-month dichotomized Glasgow Outcome Scale (GOS) score were assessed. Mean time from injury to initial CSF sampling was 10.77+/-3.13 h. On initial sampling, caffeine was detected in 24 of 30 patients, and the threshold was achieved in 9 patients. Favorable GOS was seen more often in patients with CSF caffeine concentration > or = versus < the threshold (55.6 versus 11.8%, P=0.028). Gender, age, admission CGS score, admission blood alcohol level, and admission systolic arterial blood pressure did not differ between patients with CSF caffeine concentration > or = versus < the threshold. Increases in CSF concentrations of the caffeine metabolites theobromine and paraxanthine were also associated with favorable outcome (P=0.018 and 0.056, respectively). Caffeine and its metabolites are commonly detected in CSF in patients with severe TBI and in an exploratory assessment are associated with favorable outcome. We speculate that caffeine may be neuroprotective by long-term upregulation of adenosine A1 receptors or acute inhibition of A2a receptors.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard