Merck
  • Metabolic changes in the lower esophageal sphincter influencing the result of anti-reflux surgical interventions in chronic gastroesophageal reflux disease.
Metabolic changes in the lower esophageal sphincter influencing the result of anti-reflux surgical interventions in chronic gastroesophageal reflux disease.
World journal of gastroenterology (2005-03-24)
Aron Altorjay, Arpad Juhasz, Viola Kellner, Gellert Sohar, Matyas Fekete, Istvan Sohar
ABSTRACT
With the availability of a minimally invasive approach, anti-reflux surgery has recently experienced a renaissance as a cost-effective alternative to life-long medical treatment in patients with gastroesophageal reflux disease (GERD). We are not aware of the fact whether reflux episodes causing complaints for a long time i.e., at least for one year are associated with metabolic changes in the lower esophageal sphincter, and if so, whether these may influence functional results achieved after anti-reflux surgery. Between 1 January 2001 and 31 December 2002 we performed anti-reflux surgery on 79 patients. Muscle samples were taken from the lower esophageal sphincter (LES) in 33 patients during anti-reflux surgery. Inclusion criteria were: LES resting pressure below 10 mmHg and a marked, pH proven acid exposure to the esophagus of at least one year's duration, causing subjective complaints and requiring continuous proton pump inhibitor treatment. Control samples were obtained from muscle tissue in the gastroesophageal junction that had been removed from 17 patients undergoing gastric or esophageal resection. Metabolic and lysosomal enzyme activities and special protein concentrations 16 parameters in total were evaluated in tissue taken from control specimens and tissue taken from patients with GERD. The biochemical parameters of these intra-operative biopsies were used to correlate the results of anti-reflux operations (Visick I and II-III). In the reflux-type muscle, we found a significant increase of the energy-enzyme activities e.g., creatine kinase, lactate dehydrogenase, beta-hydroxybutyrate dehydrogenase, and aspartate aminotransaminase-. The concentration of the structural protein S-100 and the myofibrillar protein troponin I were also significantly increased. Among lysosomal enzymes, we found that the activities of cathepsin B, tripeptidyl-peptidase I, dipeptidyl-peptidase II, beta-hexosaminidase B, beta-mannosidase and beta-galactosidase were significantly decreased as compared to the control LES muscles. By analyzing the activity values of the 9 patients in Visick groups II and III at two months post-surgery, we found a significant increase in the activity of the so-called energy-enzyme values and in the concentration of structural and myofibrillar proteins as compared to the rest of the reflux patients. Our results call attention to the metabolic changes that occurred in the LES muscles of reflux patients. The developing hypertrophy-like changes of LES muscles may be a reason for complaints after anti-reflux surgery, which consisted mainly of reports of persisting dysphagia.
Sigma-Aldrich
(S)-3-Hydroxybutyric acid, ≥97.0%
Pricing and availability is not currently available.