Merck
  • Evidence for axonal pathology and adaptive cortical reorganization in patients at presentation with clinically isolated syndromes suggestive of multiple sclerosis.
Evidence for axonal pathology and adaptive cortical reorganization in patients at presentation with clinically isolated syndromes suggestive of multiple sclerosis.
NeuroImage (2003-05-03)
Maria A Rocca, Domenico M Mezzapesa, Andrea Falini, Angelo Ghezzi, Vittorio Martinelli, Giuseppe Scotti, Giancarlo Comi, Massimo Filippi
ABSTRACT
Previous work has suggested that functional reorganization of cortical areas might have a role in limiting the clinical impact of axonal pathology in patients with established multiple sclerosis (MS). Since there is evidence for irreversible tissue damage even in patients with early MS, we assessed, using functional MRI (fMRI) and a general search method, the brain pattern of movement-associated cortical activations in patients at presentation with clinically isolated syndromes (CIS) suggestive of MS. To elucidate the role of cortical reorganization in these patients, we also investigated the extent to which the fMRI changes correlated with the extent of overall axonal injury of the brain. From 16 right-handed patients at presentation with CIS and 15 right-handed, age- and sex-matched healthy volunteers, we obtained: (1). fMRI (repetitive flexion-extension of the last four fingers of the right hand), (2). conventional MRI scans, and (3). a new, unlocalized proton MR spectroscopy ((1)HMRS) sequence to measure the concentration of N-acetylaspartate of the whole brain (WBNAA). Compared to controls, patients with CIS had more significant activations of the contralateral primary somatomotor cortex (SMC), secondary somatosensory cortex, and inferior frontal gyrus. They also had significant decreased WBNAA concentration. Relative activation of the contralateral primary SMC was strongly correlated with WBNAA levels (r = -0.78, P < 0.001). This study shows that axonal pathology and functional cortical changes over a rather distributed sensorimotor network occur in patients at presentation with CIS suggestive of MS and that these two aspects of the disease are strictly correlated. This suggests that the increased functional recruitment of the cortex in these patients might have an adaptive role in limiting the clinical impact of irreversible tissue damage.
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)
Pricing and availability is not currently available.