Merck
L-serine in disease and development.
The Biochemical journal (2003-01-22)
Tom J de Koning, Keith Snell, Marinus Duran, Ruud Berger, Bwee-Tien Poll-The, Robert Surtees
ABSTRACT
The amino acid L-serine, one of the so-called non-essential amino acids, plays a central role in cellular proliferation. L-Serine is the predominant source of one-carbon groups for the de novo synthesis of purine nucleotides and deoxythymidine monophosphate. It has long been recognized that, in cell cultures, L-serine is a conditional essential amino acid, because it cannot be synthesized in sufficient quantities to meet the cellular demands for its utilization. In recent years, L-serine and the products of its metabolism have been recognized not only to be essential for cell proliferation, but also to be necessary for specific functions in the central nervous system. The findings of altered levels of serine and glycine in patients with psychiatric disorders and the severe neurological abnormalities in patients with defects of L-serine synthesis underscore the importance of L-serine in brain development and function. This paper reviews these recent insights into the role of L-serine and the pathways of L-serine utilization in disease and during development, in particular of the central nervous system.
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.