Merck
  • Remarkable increase in the concentration of 8-hydroxyguanosine in cerebrospinal fluid from patients with Alzheimer's disease.
Remarkable increase in the concentration of 8-hydroxyguanosine in cerebrospinal fluid from patients with Alzheimer's disease.
Journal of neuroscience research (2002-10-23)
Takashi Abe, Hideo Tohgi, Chiaki Isobe, Takahiko Murata, Chigumi Sato
ABSTRACT
To investigate the possible role of oxidative RNA damage in the pathogenesis of Alzheimer's disease (AD), the concentrations of the oxidative stress marker 8-hydroxyguanosine (8-OHG) were measured in the cerebrospinal fluid (CSF) and the serum of patients with AD and control subjects. The concentration of 8-OHG in CSF in AD patients was approximately fivefold that in controls (P < 0.001). The concentration of 8-OHG in CSF decreased significantly with the duration of illness (r(s) = -0.48, P < 0.05) and the progression of cognitive dysfunctions (r(s) = 0.67, P < 0.01). However, the concentration of 8-OHG in CSF showed no correlation with that in serum in both the controls and AD patients. In addition, the concentration of 8-OHG in serum was not significantly altered in AD patients compared to that in controls, suggesting that the 8-OHG concentrations in the CSF do not reflect those in serum and may be probably reflect those in brain tissue. These in vivo findings suggest a possible role of 8-OHG and increased oxidative RNA damage in the early stage of the development of AD.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis