Merck
  • Accumulation of toxic products degradation of kynurenine in hemodialyzed patients.
Accumulation of toxic products degradation of kynurenine in hemodialyzed patients.
International urology and nephrology (2002-07-03)
D Pawlak, K Pawlak, J Malyszko, M Mysliwiec, W Buczko
ABSTRACT
In patients that developed a chronic renal failure the augmentation in tryptophan degradation is reflected in the increase in plasma metabolites of kynurenine pathway. Hemodialysis is one of therapeutic approaches that significantly reduce all plasma kynurenine metabolites in uremic patients. In spite of haemodialysis, plasma concentration of kynurenine, kynurenic acid, 3-hydroxykynurenine, anthranilic acid, xanthurenic acid and quinolinic acid were still elevated in uremic patients in comparison with healthy volunteers. These data shows significant disturbances in kynurenine metabolism in uremic patients. Accumulation of these substances in uremic blood is capable to account for certain uremic symptoms.
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
SAFC
L-Proline
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg