Merck
  • Ratiometric fluorescence detection of cyanide based on a hybrid coumarin-hemicyanine dye: the large emission shift and the high selectivity.
Ratiometric fluorescence detection of cyanide based on a hybrid coumarin-hemicyanine dye: the large emission shift and the high selectivity.
Chemical communications (Cambridge, England) (2011-11-03)
Xin Lv, Jing Liu, Yunlong Liu, Yun Zhao, Yuan-Qiang Sun, Pi Wang, Wei Guo
ABSTRACT
A new ratiometric fluorescent cyanide probe was developed based on the nucleophilic attack of CN(-) toward the indolium group of a hybrid coumarin-hemicyanine dye, by which high selectivity as well as large emission shift could be achieved.