Merck
  • The lethal effect of a benzamide derivative, 3-methoxybenzamide, can be suppressed by mutations within a cell division gene, ftsZ, in Bacillus subtilis.
The lethal effect of a benzamide derivative, 3-methoxybenzamide, can be suppressed by mutations within a cell division gene, ftsZ, in Bacillus subtilis.
Journal of bacteriology (1999-02-11)
Y Ohashi, Y Chijiiwa, K Suzuki, K Takahashi, H Nanamiya, T Sato, Y Hosoya, K Ochi, F Kawamura
ABSTRACT
3-Methoxybenzamide (3-MBA), which is known to be an inhibitor of ADP-ribosyltransferase, inhibits cell division in Bacillus subtilis, leading to filamentation and eventually lysis of cells. Our genetic analysis of 3-MBA-resistant mutants indicated that the primary target of the drug is the cell division system involving FtsZ function during both vegetative growth and sporulation.
Sigma-Aldrich
3-Methoxybenzamide, 97%
Pricing and availability is not currently available.