Merck
  • Mechanism of mitochondrial dysfunction and cytotoxicity induced by tropolones in isolated rat hepatocytes.
Mechanism of mitochondrial dysfunction and cytotoxicity induced by tropolones in isolated rat hepatocytes.
Chemico-biological interactions (1999-01-07)
Y Nakagawa, K Tayama
ABSTRACT
The mechanism of mitochondrial dysfunction and toxicity induced by the tropolones, beta-thujaplicin (4-isopropyl tropolone), tropolone and tropone, has been studied in freshly isolated rat hepatocytes. Incubation of hepatocytes with beta-thujaplicin (1-4 mM) elicited a concentration and time-dependent cell killing. The toxicity was accompanied by losses of cellular ATP, total adenine nucleotides and glutathione, independently of lipid peroxidation and protein thiol oxidation. The beta-thujaplicin-induced cytotoxicity was enhanced by the pretreatment of hepatocyte suspensions with EDTA (4 mM), a hydrophilic chelator, or by incubation in Ca2+ and Mg2+-deficient Krebs-Henseleit buffer. The partition coefficient of beta-thujaplicin, which formed complex with the divalent cations in Krebs-Henseleit buffer, in n-octanol/buffer was increased either in the presence of EDTA or absence of divalent cations. Comparison of toxic effects based on cell viability and adenine nucleotide levels showed that beta-thujaplicin was more toxic than tropolone or tropone in Krebs-Henseleit buffer containing EDTA (4 mM). The addition of beta-thujaplicin to isolated hepatic mitochondria reduced state 3 respiration with NAD+-linked substrate (pyruvate plus malate) and/or with an FAD-linked substrate (succinate plus rotenone), whereas state 3 respiration of ascorbate plus tetramethyl-p-phenylenediamine (cytochrome oxidase-linked respiration) was not significantly affected by beta-thujaplicin. Further, the addition of these tropolones caused a concentration-dependent increase in the rate of state 4 oxygen consumption, indicating an uncoupling effect. These results indicate that beta-thujaplicin- and tropolone-induced cytotoxicity are associated with an acute ATP depletion via mitochondrial dysfunction related to oxidative phosphorylation and that the induction of cytotoxicity is affected by EDTA or divalent cations.
Sigma-Aldrich
Tropone, 97%
Pricing and availability is not currently available.