Merck
  • HPLC on Chiralcel OJ-R for enantiomer separation and analysis of ketoprofen, from horse plasma, as the 9-aminophenanthrene derivative.
HPLC on Chiralcel OJ-R for enantiomer separation and analysis of ketoprofen, from horse plasma, as the 9-aminophenanthrene derivative.
The Journal of pharmacy and pharmacology (1998-05-26)
H Y Aboul-Enein, A Van Overbeke, G Vander Weken, W Baeyens, H Oda, P Deprez, A De Kruif
ABSTRACT
Racemic ketoprofen is a non-steroidal anti-inflammatory drug used to treat musculoskeletal and colic conditions in horses. The enantioselective chiral inversion of ketoprofen administered to horses has been studied by use of cellulose tris(4-methylbenzoate), also known as Chiralcel OJ-R, as chiral stationary phase; acetonitrile - 0.02 M perchlorate buffer (pH 2.0)-methanol, 60:15:25 (v/v/v) was used as mobile phase. Before chromatography, to effect adequate chiral interaction with the chiral stationary phase ketoprofen was derivatized with 9-aminophenanthrene, under acid conditions, after solid-phase (C18) extraction and then liquid-liquid extraction, to ensure effective removal of endogenous plasma materials. The 9-aminophenanthrene derivative of S-ibuprofen was used as internal standard. The enantiomers of ketoprofen were separated to baseline (Rs = 6.44, alpha = 1.76) within a short analysis time. The results indicate that the bio-inversion of R-ketoprofen to the S isomer is significant in equine species. However, considerable differences in pharmacokinetic parameters were observed, indicating large inter-animal variation.
Sigma-Aldrich
9-Aminophenanthrene, 96%
Pricing and availability is not currently available.