Merck
  • Enzymatic elimination of heparin from plasma for activated partial thromboplastin time and prothrombin time testing.
Enzymatic elimination of heparin from plasma for activated partial thromboplastin time and prothrombin time testing.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (1993-08-01)
A M van den Besselaar, J Meeuwisse-Braun
ABSTRACT
The prolongation of the APTT and PT by heparin can be neutralized by treating the plasma with heparinase or with certain anion-exchangers. The purpose of the present study was to assess the effectiveness of a new commercial heparinase reagent. Heparinase (Hepzyme) for the removal of heparin from plasma was compared with ECTEOLA-cellulose treatment. In ex vivo heparin samples, shorter APTT values were obtained with heparinase than with ECTEOLA-cellulose, suggesting that the former treatment is more effective for the removal of heparin from plasma. Heparinase (Hepzyme) treatment may be used to improve INR assessment in patients treated with heparin and oral anticoagulants concomitantly.
Sigma-Aldrich
Ecteola Cellulose, coarse
Pricing and availability is not currently available.