Merck
  • Quantitative capillary column gas chromatographic method for the determination of glycopyrronium in human plasma.
Quantitative capillary column gas chromatographic method for the determination of glycopyrronium in human plasma.
Journal of chromatography (1984-06-08)
G R Murray, T N Calvey, N E Williams, K Chan
ABSTRACT
A new sensitive and selective capillary column gas chromatographic method for the anti-cholinergic agent glycopyrronium bromide in human plasma is described. The procedure involves preliminary ion-pair extraction of the drug into dichloromethane, followed by concentration and analysis of the ion-pair complex by capillary column gas chromatography using a nitrogen-sensitive detector. The method depends on the thermal dequaternisation of the quaternary ammonium compound and can be used to detect 5 ng/ml in a 3-ml plasma sample. The assay procedure has been applied to the determination of the plasma concentration of glycopyrronium after intravenous administration to an anaesthetised patient.
Product Description
Supelco
Mepenzolate bromide, analytical standard
Pricing and availability is not currently available.