Merck
  • Platelet adhesion onto microcapsule membranes with different degrees of sulfonation.
Platelet adhesion onto microcapsule membranes with different degrees of sulfonation.
Journal of biomedical materials research (1983-11-01)
N Muramatsu, T Kondo
ABSTRACT
Microcapsules having different numbers of sulfonic acid groups on their surfaces were prepared by the interfacial polycondensation method. Platelets adhered to these capsules, and the rate and degree of platelet adhesion were found to be remarkable on those microcapsules that had high surface charge compared to those with low surface charge. This trend was strengthened by the coating of the capsules with plasma, while the electrophoretic mobilities of the plasma-coated capsules showed a considerable reduction and no appreciable difference between the respective mobilities could be observed. This trend suggest that the adsorption of plasma components on microcapsules, and not their surface negative charge, affected platelet adhesiveness directly. In connection with the glycosyl transferase hypothesis, the adsorption pattern and its effect on platelet adhesion of albumin, gamma-globulin, and fibrinogen were examined. The protein adsorption pattern varied depending on both the type of protein and the magnitude of negative charge on the microcapsule surface, but its effect on platelet adhesion was not fully consistent with the glycosyl transferase hypothesis. It was concluded from the findings that the surface negative charge of the capsules strongly affected the composition, molecular orientation, and/or configuration of the adsorbed plasma components, which probably induced the differences in platelet adhesiveness depending on the magnitude of surface negative charge of the microcapsules.
Sigma-Aldrich
4,4′-Diamino-2,2′-stilbenedisulfonic acid, technical grade, 85%