Merck
Allergic contact dermatitis due to an epoxy acrylate.
The British journal of dermatology (1981-06-01)
J R Nethercott
ABSTRACT
A printer developed allergic contact dermatitis due to epoxy acrylate resin. Incomplete cross-reactivity between four epoxy acrylate resins and a lack of cross-reactivity to epoxy resin was demonstrated. The epoxy acrylate resin has been shown to be a potent cutaneous sensitizer for the guinea-pig.
Sigma-Aldrich
Bisphenol A glycerolate (1 glycerol/phenol) diacrylate, contains MEHQ as inhibitor