Merck
[Modified histological methods using a nuclear dye gallocyanine].
Arkhiv patologii (1982-01-01)
V V Zadorozhnyĭ
ABSTRACT
It is suggested that a nuclear stain; gallocyanine, be used simultaneously with the three main histological staining methods: hematoxylin-eosin, Van-Gieson's, and Nissl' stainings. This modification is technically simple, gives preparations of a high quality, and is time- and labour-saving.
Sigma-Aldrich
Gallocyanine, Dye content 90 %