Merck
  • Liquid chromatographic determination of 2,4-dinitroaniline and 2-naphthol in D&C Orange No. 17.
Liquid chromatographic determination of 2,4-dinitroaniline and 2-naphthol in D&C Orange No. 17.
Journal - Association of Official Analytical Chemists (1985-05-01)
A L Scher
ABSTRACT
A method is described for the determination of the intermediates in D&C Orange No. 17 by reverse phase liquid chromatography. The pigment is dissolved in boiling dioxane and then precipitated. The filtrate is chromatographed by isocratic elution, which is followed by a wash and equilibration. Peak area calibrations were linear. At the provisional specification levels, 99% prediction limits were 0.200 +/- 0.0012% 2,4-dinitroaniline (2,4-DNA) and 0.200 +/- 0.006% 2-naphthol. The limits of determination were 0.0023% for 2,4-DNA and 0.013% for 2-naphthol at the 99.5% confidence level. Recoveries were 98-100% for 2,4-DNA added at the 0.005-2% level, and 93-103% for 2-naphthol added at the 0.025-2% level. A survey of certified D&C Orange No. 17 samples showed that the lots contained higher levels of the intermediates than were determined previously by a cellulose column method, in which the pigment is not dissolved.
Product Description
Supelco
Pigment Orange 5, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2,4-Dinitroaniline, 98%
Pricing and availability is not currently available.