- Ineffective treatment of essential tremor with an alcohol, methylpentynol.
Ineffective treatment of essential tremor with an alcohol, methylpentynol.
Journal of neurology, neurosurgery, and psychiatry (1986-02-01)
H Teräväinen, J Huttunen, P Lewitt
PMID3512778
ABSTRACT
Six patients with essential tremor tested in the therapeutic effectiveness of a 6-carbon alcohol, methylpentynol, 200 mg/day, against placebo in a randomised double-blind clinical cross-over trial. The effect of methylpentynol on postural tremor amplitude was not different from that of placebo.