- Acetylation, deacetylation and hydroxylation of sulphamethoxazole and N4-acetylsulphamethoxazole in the snail Cepaea hortensis.
Acetylation, deacetylation and hydroxylation of sulphamethoxazole and N4-acetylsulphamethoxazole in the snail Cepaea hortensis.
The Veterinary quarterly (1987-10-01)
T B Vree, M L Vree, J F Nouws
PMID3424644
ABSTRACT
The snail Cepaea hortensis is able to acetylate and hydroxylate sulphamethoxazole in a similar way to man. About one percent deacetylation, but no hydroxylation of N4-acetysulphamethoxazole takes place. The relative rates of the metabolic pathways for hydroxylation and acetylation of sulphamethoxazole are equal.
Product Description
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard