Merck
  • Strong absorption and selective emission from engineered metals with dielectric coatings.
Strong absorption and selective emission from engineered metals with dielectric coatings.
Optics express (2013-04-11)
W Streyer, S Law, G Rooney, T Jacobs, D Wasserman
ABSTRACT
We demonstrate strong-to-perfect absorption across a wide range of mid-infrared wavelengths (5-12µm) using a two-layer system consisting of heavily-doped silicon and a thin high-index germanium dielectric layer. We demonstrate spectral control of the absorption resonance by varying the thickness of the dielectric layer. The absorption resonance is shown to be largely polarization-independent and angle-invariant. Upon heating, we observe selective thermal emission from our materials. Experimental data is compared to an analytical model of our structures with strong agreement.
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis