Merck
  • Palladium-catalyzed desulfitative cross-coupling reaction of sodium sulfinates with benzyl chlorides.
Palladium-catalyzed desulfitative cross-coupling reaction of sodium sulfinates with benzyl chlorides.
Organic letters (2013-04-06)
Feng Zhao, Qi Tan, Fuhong Xiao, Shufeng Zhang, Guo-Jun Deng
ABSTRACT
A palladium-catalyzed approach for the synthesis of diarylmethanes from sodium sulfinates and benzyl chlorides is described. Various aromatic sodium sulfinates were used as aryl sources via extrusion of SO2 and gave the diarylmethanes in moderate to good yields.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis