Merck
  • Structures of (1→6)-β-D-glucans from Bulgaria inquinans (Fries) and their immunological activities.
Structures of (1→6)-β-D-glucans from Bulgaria inquinans (Fries) and their immunological activities.
Carbohydrate polymers (2013-03-19)
Hongtao Bi, Tingting Gao, Dongbo Liu, Guihua Tai, Min Wei, Yifa Zhou
ABSTRACT
In previous study, an unbranched (1→6)-β-D-glucan with Mw 2.6kDa was isolated from fruit bodies of Bulgaria inquinans (Fries). In present paper, three branched (1→6)-β-D-glucans were obtained from the water-extracted residues by a sequential KOH-extraction, namely BIK2, BIK10 and BIK30. Their molecular weights were determined to be 37.5kDa (BIK2), 288.9kDa (BIK10) and 175.5kDa (BIK30). Structural analysis indicated that their backbones were substituted by single glucosyls at C-3 positions, the branching ratios were 0.01 (BIK2), 0.17 (BIK10), 0.25 (BIK30). Immunological tests showed that all the four β-D-glucans could significantly increase the ConA or LPS-induced lymphocytes proliferation in vivo. Moreover, branched (1→6)-β-D-glucans have more significantly lymphocytes proliferation activities than unbranched (1→6)-β-D-glucan, and the effect of (1→6)-β-D-glucans on lymphocytes proliferation increases along with molecular weights. The present results well enrich the structure-activity relationships of (1→6)-β-D-glucan, and indicate (1→6)-β-D-glucans from B. inquinans (Fries) are potential immunostimulating agents.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets